Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium osmate(VI) dihydrate

Catalog No. 20910425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-910-425G 2.5 g
20-910-4100MG 100 mg
20-910-4500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-910-425G Supplier Sigma Aldrich Supplier No. 2091042.5G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the oxidative cleavage of olefins to ketones and carboxylic acids. Also employed in an intramolecular amidohydroxylation of carbamoyloxy-tethered olefins providing hydroxy oxazolidinones.

Specifications

CAS 10022-66-9
Quantity 2.5 g
Molecular Formula K2OsO4 -+ 2H2O
Linear Formula K2OsO4-+2H2O
MDL Number MFCD00149919
Molecular Weight (g/mol) 368.45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.