Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Oxalate, 30% (w/v), Ricca Chemical
SDP

Catalog No. R633000020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R633000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R633000020F Supplier Ricca Chemical Company Supplier No. R633000020F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium hydrate oxalate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Assay Percent Range 30% (w/v)
Percent Purity 30% W/V
Grade Laboratory
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Potassium Oxalate, Aqueous Solution
Recommended Storage 15°C to 30°C (59°F to 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.