Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Oxalate, 20% (w/v), Ricca Chemical
SDP

Catalog No. R63250004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R63250004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R63250004A Supplier Ricca Chemical Company Supplier No. R63250004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 6487-48-5
Molecular Weight (g/mol) 166.22 g/mol
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Physical Form Solution
Quantity 4 L
Assay Percent Range 20% (w/v)
Solubility Information Solubility in Water: Infinite
Percent Purity 20% W/V
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Appearance Clear colorless liquid
Chemical Name or Material Potassium Oxalate
Recommended Storage Health
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.