Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Oxalate, For Dissolved Oxygen, Certified, 2.0% (w/v) ±0.1%, LabChem™

Catalog No. LC198401 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC198401 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC198401 Supplier LabChem, Inc. Supplier No. LC198401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aqueous potassium oxalate solution for use in dissolved oxygen analysis.

Potassium Oxalate 2% w/v solution, prepared for dissolved oxygen analysis.

  • Compendia References: APHA 4500-O D, ASTM D888-09 A
  • Traceable to NIST: Yes
  • Composition: 98% Water, 2% Potassium Oxalate, Monohydrate, ACS
  • Physical State: Liquid
  • Specific Gravity: 1 g/mL
  • Stability: Stable under normal conditions
  • Storage: Keep container closed when not in use

Specifications

Color Clear, colorless
Volume (Metric) 500 mL
Format Liquid
Packaging Poly Bottle
Quantity 500 mL
Storage Requirements Room Temperature
Type Potassium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.