Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Oxalate Monohydrate
SCI_ED

Catalog No. S25496 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25496 100 g Lab Grade
S25496A 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25496
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium hydrate oxalate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Color White
Density 2.13g/mL
Assay Percent Range >99%
Packaging Lab Grade
Linear Formula K2C2O4·H2O
Quantity 100 g
Formula Weight 184.24
Grade Laboratory
Chemical Name or Material Potassium Oxalate, Monohydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.