Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Oxalate Monohydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. P273500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P273-500 500 g Glass Bottle
P273-250 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P273-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium hydrate oxalate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 356°C
Color White
Density 2.13g/cm³
pH 7.0 to 8.5
Assay Percent Range 98.5 to 101.0%
Packaging Glass Bottle
Quantity 500 g
UN Number 2811
Solubility Information 0.01% max. Insoluble Matter
Formula Weight 184.23
Percent Purity 98.5 to 101.0%
Grade Certified ACS
Physical Form Solid
Chemical Name or Material Potassium Oxalate Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.