Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium oxalate monohydrate, Honeywell™

Catalog No. 6002777 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg

Purum p.a.,≥99.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium;oxalate;hydrate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Quantity 250 g
Linear Formula (COOK)2 · H2O
UN Number NONH for all modes of transport
Beilstein 3752576
Formula Weight 184.23g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Potassium oxalate monohydrate
Hazard Statement P280-P301 + P312 + P330
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.