Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Potassium oxonate | 2207-75-2 | 99.5% | 195.17 | 10 G
SDP

Catalog No. 5000386571
Encompass_Preferred

Potassium oxonate (Potassium azaorotate) is a uricase inhibitor that inhibits the phosphorylation of 5-FU to 5-fluorouridine-5'-monophosphate.

  • Target: Xanthine oxidase
  • Research areas: Metabolic disease
  • CAS number: 2207-75-2
  • Molecular formula: C4H2KN3O4
  • Molecular weight: 195.17
  • Purity: 99.52%
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Catalog No. 50-003-86571 Supplier Medchemexpress LLC Supplier No. HY1751110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potassium oxonate (Potassium azaorotate) is a uricase inhibitor that inhibits the phosphorylation of 5-FU to 5-fluorouridine-5'-monophosphate.

  • Target: Xanthine oxidase
  • Research areas: Metabolic disease
  • CAS number: 2207-75-2
  • Molecular formula: C4H2KN3O4
  • Molecular weight: 195.17
  • Purity: 99.52%
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.