Learn More
Selleck Chemical LLC Potassium oxonate-E0353-1G
SDP
Supplier:  Selleck Chemical LLC E03531G
Encompass_Preferred
Potassium oxonate (Oxonic Acid Potassium Salt) is a selectively competitive uricase inhibitor produced hyperuricemia (HUA) in rodents
Catalog No. 50-313-8364