Learn More
Medchemexpress LLC Potassium p-nitrophenyl sulfate | 6217-68-1 | 99.9% | 257.26 | 1 G
SDP
Catalog No. 5000366905
Encompass_Preferred
p-Nitrophenyl Sulfate Potassium is a chromogenic substrate for arylsulfatase. It is released by arylsulfonase cleavage, and the activity of arylsulfonase can be quantitatively determined by colorimetric detection at 400 nm.
  • Chromogenic substrate for arylsulfatase
  • Useful for quantitative determination of arylsulfonase activity
  • Colorimetric detection at 400 nm
Catalog No. 50-003-66905 Supplier Medchemexpress LLC Supplier No. HYW0094541G
p-Nitrophenyl Sulfate Potassium is a chromogenic substrate for arylsulfatase. It is released by arylsulfonase cleavage, and the activity of arylsulfonase can be quantitatively determined by colorimetric detection at 400 nm.
  • Chromogenic substrate for arylsulfatase
  • Useful for quantitative determination of arylsulfonase activity
  • Colorimetric detection at 400 nm