Learn More
Medchemexpress LLC Potassium p-tolyl sulfate | 91978-69-7 | MFCD28100835 | >98.0% | 226.29 g/mol | C7H7KO4S | 5 MG
SDP
Catalog No. 5000263148
Encompass_Preferred
p-Cresyl sulfate potassium is the potassium salt form of p-cresyl sulfate supplied as an analytical standard for research use. It is provided for use as a reference material in biochemical, toxicology, and analytical method development to support studies of uremic toxins and related metabolic pathways.
  • Analytical standard for quantitative and qualitative assays.
  • Used in biochemical and toxicology studies of uremic toxins.
  • Reported purity >98.0% (HPLC).
  • Chemical formula C7H7KO4S; molecular weight 226.29 g/mol.
  • Supplied as a 5 MG powder for laboratory use.
Catalog No. 50-002-63148 Supplier Medchemexpress LLC Supplier No. HY111431AR5MG
p-Cresyl sulfate potassium is the potassium salt form of p-cresyl sulfate supplied as an analytical standard for research use. It is provided for use as a reference material in biochemical, toxicology, and analytical method development to support studies of uremic toxins and related metabolic pathways.
  • Analytical standard for quantitative and qualitative assays.
  • Used in biochemical and toxicology studies of uremic toxins.
  • Reported purity >98.0% (HPLC).
  • Chemical formula C7H7KO4S; molecular weight 226.29 g/mol.
  • Supplied as a 5 MG powder for laboratory use.