Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium P-Tolyltrifluoroborate

Catalog No. 5715551G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-155-51G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 57-155-51G Supplier Sigma Aldrich Supplier No. 5715551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassiump-tolyltrifluoroborate can be used:As a precursor/starting material for the synthesis of biaryl compounds by reacting with various aryl halides using Pd/C catalyst.As a reagent in the carbonylative arylation of vinyl ketones via 1,4-addition.As a substrate in the synthesis of primary arylamines by reacting with hydroxylamine-O-sulfonic acid under metal-free conditions.

Specifications

CAS 761446-44-0
Melting Point 59°C to 64°C
Molecular Formula C10H17BN2O2
Linear Formula C10H17BN2O2
MDL Number MFCD03789259
Quantity 5 g
Synonym 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole; 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole; 1-Methyl-4-pyrazoleboronic acid pinacol ester; 2-(1-Methylpyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-methylpyrazole
Molecular Weight (g/mol) 208.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.