Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium perchlorate, 99+%, for analysis

Catalog No. ac221360250
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221360250 25 g Plastic bottle
AC221365000 500 g Plastic bottle
AC221360010 1 kg Plastic bottle
AC221360025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC221360250 Supplier Thermo Scientific Chemicals Supplier No. 221360250
Only null left
Add to Cart

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-74-7
Molecular Formula ClKO4
Molecular Weight (g/mol) 138.54
MDL Number MFCD00011362
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
PubChem CID 516900
IUPAC Name potassium;perchlorate
SMILES [K+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 400.0°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99+%
Linear Formula KClO4
Merck Index 15,7774
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol
Formula Weight 138.55
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium perchlorate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-912-9
RTECSNumber SC9700000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.