Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.0420 N, Ricca Tightrant™
SDP

Catalog No. R63860001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R63860001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R63860001C Supplier Ricca Chemical Company Supplier No. R63860001C
 Restricted Item
Add to Cart
Add to Cart
  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Specifications

Chemical Name or Material Potassium Permanganate
Color Purple Liquid
CAS 7732-18-5, 7722-64-7
CAS Max % 99.86, 0.14
Assay Percent Notes Normal at 20°C
Concentration 0.04199 to 0.04201 N
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.