Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.100 N (N/10), 0.0200 M (M/50), Ricca Tightrant™
SDP

Catalog No. R6400000200 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6400000200 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6400000200 Supplier Ricca Chemical Company Supplier No. 6400200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Specifications

Color Colorless
CAS 7722-64-7, 7732-18-5
Odor Odorless
Packaging Amber Glass Bottle
Assay Percent Range 0.1000 ± 0.0001 N at 20°C
Solvent or Matrix Aqueous Solution
Molecular Formula KMnO4
Quantity 200 mL
Physical Form Liquid
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
Solubility Information Solubility in Water: Infinite
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
PubChem CID 516875
Grade Laboratory
Chemical Name or Material Potassium Permanganate, 0.1N
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.