Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium permanganate, 0.1N Standardized Solution

Catalog No. AA35590K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35590K2 1 L
AA35590K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35590K2 Supplier Thermo Scientific Chemicals Supplier No. 035590K2
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol

Potassium permanganate, 0.1N standardized solution is used for volumetric analysis in laboratories. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium permanganate
Color Purple
CAS 7722-64-7
Physical Form Liquid
Quantity 1 L
Molecular Formula KMnO4
MDL Number MFCD00011364
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
Solubility Information Soluble in water.
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
PubChem CID 516875
Formula Weight 158.03
Sensitivity Light sensitive
Appearance Dark purple liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.