Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.63% (w/v), Ricca Chemical
SDP

Catalog No. 635532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6355-32 1 L Amber Glass Bottle
635516 500 mL Amber Glass Bottle
6355-1 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6355-32 Supplier Ricca Chemical Company Supplier No. 635532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For ASTM Testing

Specifications

CAS 7732-18-5
Molecular Weight (g/mol) 158.03 g/mol
Percent Purity 0.63%
Grade Laboratory
Packaging Amber Glass Bottle
Methods ASTM for Dissolved Oxygen (DO)
Quantity 1 L
Chemical Name or Material Potassium Permanganate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.