Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Permanganate
SCI_ED

Catalog No. S25497 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25497 100 g Reagent Grade
S25497A 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25497
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Brown
Quantity 100 g
Assay Percent Range >95%
Linear Formula KMnO4
Formula Weight 158.04
Packaging Reagent Grade
Chemical Name or Material Potassium Permanganate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.