Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate
SCI_ED

Catalog No. S77642 Shop All Fisherbrand Products
Change view
Click to view available options
Reagent Grade - Crystal; 100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S77642 Reagent Grade - Crystal; 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S77642

Specifications

Color Purple
CAS 7722-64-7
Quantity 100 g
Molecular Formula KMnO4
Linear Formula KMnO4
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
PubChem CID 516875
Formula Weight 158.04
Packaging Reagent Grade - Crystal
Chemical Name or Material Potassium Permanganate
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife Indefinite
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.