Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium permanganate, 98%

Catalog No. AAA1217036
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1217036 500 g
AAA121700E 2500 g
AAA121700C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1217036 Supplier Thermo Scientific Chemicals Supplier No. A1217036
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol

Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

Solubility
Soluble in water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, peroxides, zinc, copper, alcohols, hydrogen fluoride and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Melting Point 240°C (decomposition)
Quantity 500 g
UN Number UN1490
Merck Index 14,7655
Solubility Information Soluble in water.
Formula Weight 158.03
Percent Purity 98%
Sensitivity Light sensitive
Density 2.7
Chemical Name or Material Potassium permanganate
Hazard Category H272-H302-H314-H335-H361d-H373
Hazard Statement GHS H Statement
H272-H302
May intensify fire
oxidizer.
Harmful if swallowed.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM PERMANGANATE
EINECSNumber 231-760-3
RTECSNumber SD6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.