Learn More
Potassium permanganate, ACS, 99.0% min
Description
In photography, tanning leather, purifying water, manufacturing mineral water, as an analytical lab reagentPotassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 240°C (decomposition) |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Sensitivity | Light sensitive |
| Appearance | Dark crystalline |
| Decomposition Information | Decomposes in organic solvents and acids. |
| Assay Percent Range | 99.0% min. |
| Density | 2.7 g/cm3 |
| Solubility Information | Partially soluble in water. Decomposes in organic solvents and acids. |
| Formula Weight | 158.03 |
| Show More |
Safety and Handling
| Hazard Category | H272-H302-H314-H335-H361d-H373 |
| Hazard Statement | This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1, Packing Group: II |
| EINECSNumber | 231-760-3 |
| RTECSNumber | SD6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.