Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, ACS Grade, ≥ 99.0 %, LabChem™

Catalog No. LC198501 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC198501 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC198501 Supplier LabChem, Inc. Supplier No. LC198501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Melting Point 240°C
Color Purple
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula KMnO4
UN Number UN1490
Solubility Information Soluble in water
Formula Weight 158.04
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2700kg/m3
Decomposition Information Manganese
Density 2,700kg/m3
Chemical Name or Material Potassium Permanganate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective clothing, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-760-3
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.