Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium permanganate ACS25KG
Catalog No. 502899204
Encompass
Potassium permanganate a dark violet colored crystalline solid is a strong oxidizing agent for a wide range of organic compounds. It is commonly used as a disinfectant to treat external infections in aquaculture. Corncobs pretreated with alkaline potassium permanganate SOLUTIONon showed high carbohydrate recovery due to the increased enzymatic hydrolysis of lignocellulose. The acidic potassium permanganate acts a chemiluminescence reagent for the determination of organic compounds.
Catalog No. 50-289-9204 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2234682.5KG
Potassium permanganate a dark violet colored crystalline solid is a strong oxidizing agent for a wide range of organic compounds. It is commonly used as a disinfectant to treat external infections in aquaculture. Corncobs pretreated with alkaline potassium permanganate SOLUTIONon showed high carbohydrate recovery due to the increased enzymatic hydrolysis of lignocellulose. The acidic potassium permanganate acts a chemiluminescence reagent for the determination of organic compounds.