Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, Certified, 0.0500N ±0.0005N (0.01M), LabChem™

Catalog No. LC199604 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC199604 4 L
LC199602 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC199604 Supplier LabChem, Inc. Supplier No. LC199604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Purple
CAS Max % 99.84, 0.16
Physical Form Liquid
Quantity 4 L
Linear Formula KMnO4
Solubility Information Soluble in water
Formula Weight 158.04
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0500N ±0.0005N (0.01M)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Decomposition Information Manganese; Oxygen
Density 1g/mL
Chemical Name or Material Potassium Permanganate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.