Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, For Chromium, 4% (w/v), LabChem™

Catalog No. LC199302 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC199302 1 L
LC199301 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC199302 Supplier LabChem, Inc. Supplier No. LC199302
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Purple
CAS Max % 96, 4
Linear Formula KMnO4
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 158.04
Percent Purity 4% (w/v)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Decomposition Information Manganese; Oxygen
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Permanganate, For Chromium
Show More Show Less
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.