Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, Purified, Reagents

Catalog No. 01337094
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-094 2.5 kg
01-337-093 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-094 Supplier Reagents Holdings Llc Supplier No. C22885002.5A5
Add to Cart
Add to Cart

Potassium Permanganate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.03
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
IUPAC Name potassium manganesoylolate
SMILES [K+].[O-][Mn](=O)(=O)=O

Specifications

Packaging Poly Bottle
Specific Gravity 2.7
Physical Form Solid
Color Purple/Black
Melting Point >240°C
Quantity 2.5 kg
Chemical Name or Material Potassium Permanganate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.