Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate Solution, 0.5%, ENG Scientific

Catalog No. ES6971
Change view
Click to view available options
Quantity:
2 x 250 mL
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6971 1 gal.
ES6970 2 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES6971 Supplier Eng Scientific Inc Supplier No. 6971
Add to Cart
Add to Cart

ENG Scientific Potassium Permanganate Solution, 0.5% uses a phenolated fluorescent dye in place of carbol fuchsin.

  • Flourescent Stain Kit for acid-fast bacteria
  • Potassium Permanganate 0.5%, Solution III

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.03
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium manganesoylolate
SMILES [K+].[O-][Mn](=O)(=O)=O

Specifications

Packaging Bottle
Quantity 1 gal.
Chemical Name or Material Potassium Permanganate Solution

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.