Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate Solution, Fisher Chemical™
SDP

Catalog No. SP9700500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP9700-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SP9700-500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7722-64-7, 7732-18-5
CAS Min % 93.0, 4.67
CAS Max % 97.0, 4.86
Packaging Amber Glass Bottle
Methods For AOAC Method 932.05
Molecular Formula KMnO4
Quantity 500 mL
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
PubChem CID 516875
Percent Purity 5%
Grade Laboratory
Chemical Name or Material Potassium Permanganate Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.