Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium permanganate solution, Volumetric, 0.2 M KMnO4 (1N), Solstice

Catalog No. 60046539 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-539 6 x 1 L
60-012-96 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-539 Supplier Solstice Supplier No. 351846X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.2 M KMnO4 (1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%
Physical Form Liquid
Quantity 6 x 1 L
Linear Formula KMnO4
UN Number UN3082
Formula Weight 158.03
Grade Not Available
Concentration or Composition (by Analyte or Components) 0.2 M KMnO4 (1N)
Packaging Glass Bottle
Chemical Name or Material Potassium permanganate solution, Volumetric, 0.2 M KMnO4 (1N)
Show More Show Less
Hazard Statement P273-P501
ShelfLife 540 days from date of manufacture
DOTInformation 9 / PGIII
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.