Learn More
Strem, An Ascensus Company CAS# 10378-50-4. 1g. Potassium perruthenate, 98%. MFCD00061333
SDP
Catalog No. 5090111969
Encompass
CAS# 10378-50-4. 1g. Potassium perruthenate, 98%. MFCD00061333. Molecular Weight: 204.17. Molecular Formula: KRuO4. Color/form: black xtl. Strem# 44-1750. http://www.strem.com/catalog/v/44-1750/
Catalog No. 50-901-11969 Supplier Strem, An Ascensus Company Supplier No. 4417501G
CAS# 10378-50-4. 1g. Potassium perruthenate, 98%. MFCD00061333. Molecular Weight: 204.17. Molecular Formula: KRuO4. Color/form: black xtl. Strem# 44-1750. http://www.strem.com/catalog/v/44-1750/