Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Persulfate, 5% (w/v), Ricca Chemical
SDP

Catalog No. 645016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6450-16 500 mL
6450-32 1 L
R64500004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6450-16 Supplier Ricca Chemical Company Supplier No. 645016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7727-21-1
Molecular Formula K2O8S2
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
PubChem CID 24412

Specifications

Physical Form Liquid
Quantity 500 mL
Packaging Natural Poly Bottle
Synonym potassium peroxydisulfate, potassium peroxodisulfate, potassium peroxydisulphate
SMILES [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
IUPAC Name dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Weight (g/mol) 270.31
Percent Purity 5% W/V
Grade Laboratory
Chemical Name or Material Potassium Persulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.