Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Persulfate ACS AR, Macron Fine Chemicals™

Catalog No. MK7076500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7076-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7076-500 Supplier Avantor Supplier No. 707612
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7727-21-1
Quantity 500 g
Molecular Formula K2O8S2
Synonym potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;sulfonatooxy sulfate
Molecular Weight (g/mol) 270.309
PubChem CID 24412
Formula Weight 270.32
Percent Purity ≥99%
Grade ACS
Packaging Glass
Chemical Name or Material Potassium Persulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.