Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), Reagents

Catalog No. 01337804
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-804 500 g
01-337-806 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-804 Supplier Reagents Holdings Llc Supplier No. C1288600500A5
Add to Cart
Add to Cart

Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.31
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym Potassium Peroxydisulfate, Potassium Peroxodisulphate
IUPAC Name dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point >170°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.477
Chemical Name or Material Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.