Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Persulfate, Solstice

Catalog No. 6001309 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-09 100 g
60-013-11 5 g
60-013-10 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-013-09 Supplier Solstice Supplier No. 216224100G

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.309
MDL Number MFCD00011386
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
PubChem CID 24412
IUPAC Name dipotassium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Specifications

Quantity 100 g
Linear Formula K2S2O8
UN Number 1492
Formula Weight 270.32g/mol
Percent Purity ≥99%
Grade ACS Reagent
Density Vapor Density: 9.3 (vs air)
Chemical Name or Material Potassium Persulfate
Hazard Statement P220-P261-P280-P305 + P351 + P338-P342 + P311
Precautionary Statement H272-H302-H315-H317-H319-H334-H335
EINECSNumber 231-781-8
RTECSNumber SE0400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.