Learn More
Hach Company Potassium Persulfate Powder Pillows, pk/100
Supplier:  Hach Company 245199
Encompass
For determination of total phosphate by acid persulfate digestion. Hach method 8190. Pack of 100 powder pillows.
Catalog No. 50-199-4776