Learn More
Hach Company Potassium Persulfate Powder Pillows, pk/100
Supplier:  Hach Company 2084769
Encompass
For total phosphate determination by acid persulfate digestion and phosphonate determination by the UV Oxidation method. Pack of 100 powder pillows.
Catalog No. 50-199-5254