Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium phosphate, 0.2M buffer soln., pH 7.6, Thermo Scientific™

Catalog No. AAJ63128AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63128AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63128AP Supplier Thermo Scientific Chemicals Supplier No. J63128AP

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble to slightly soluble in ethanol, insoluble in alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7778-77-0
Concentration 0.2 M
pH 7.6
Physical Form Liquid
Quantity 500 mL
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.