Learn More
Potassium phosphate, anhydrous, 97%
Description
Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1340°C |
| Physical Form | Granules |
| Quantity | 1 kg |
| Assay Percent Range | 97% |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 212.27 |
| Sensitivity | Hygroscopic |
| Density | 2.564 g/mL |
| Chemical Name or Material | Potassium phosphate, anhydrous |
Safety and Handling
| Hazard Category | H318-H335 |
| Precautionary Statement | P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c |
| EINECSNumber | 231-907-1 |
| RTECSNumber | TC8450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.