Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium phosphate dibasic trihydrate for molecular biology, >=99% | 16788-57-1 | MFCD00149926 | 1KG
Catalog No. NC1475072
Encompass
Potassium phosphate dibasic trihydrate for molecular biology, >=99% | Purity: >=99% | Mol Wt: 228.22 | 16788-57-1 | MFCD00149926 | 1KG
Catalog No. NC1475072 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P96661KG
Potassium phosphate dibasic trihydrate for molecular biology, >=99% | Purity: >=99% | Mol Wt: 228.22 | 16788-57-1 | MFCD00149926 | 1KG