Learn More
Medchemexpress LLC Potassium phosphate, dibasic | 7758-11-4 | MFCD00011383 | 100.0% | 174.17 g/mol | K2HPO4 | 250 G
SDP
Catalog No. 5000303725
Encompass_Preferred
Dipotassium hydrogen phosphate (potassium phosphate dibasic, K2HPO4) is an anhydrous, white crystalline salt used as a buffering agent, pH adjuster, and pharmaceutical excipient. It offers reliable buffering capacity in aqueous formulations and is suitable for laboratory and formulation applications where high purity is required.
  • Provides stable buffering capacity in aqueous systems.
  • High purity suitable for analytical and formulation use.
  • Soluble in water; may require sonication for complete dissolution.
  • Available in multiple laboratory-scale pack sizes.
  • Useful for pH control and excipient formulations.
Catalog No. 50-003-03725 Supplier Medchemexpress LLC Supplier No. 250GHYW019883
Dipotassium hydrogen phosphate (potassium phosphate dibasic, K2HPO4) is an anhydrous, white crystalline salt used as a buffering agent, pH adjuster, and pharmaceutical excipient. It offers reliable buffering capacity in aqueous formulations and is suitable for laboratory and formulation applications where high purity is required.
  • Provides stable buffering capacity in aqueous systems.
  • High purity suitable for analytical and formulation use.
  • Soluble in water; may require sonication for complete dissolution.
  • Available in multiple laboratory-scale pack sizes.
  • Useful for pH control and excipient formulations.