Learn More
Potassium Phosphate Dibasic, 99%, (Crystalline), MP Biomedicals
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Physical Form | Crystalline |
| Quantity | 5 kg |
| Assay Percent Range | 99% |
| Formula Weight | 174.2 |
| Percent Purity | 100% |
| Chemical Name or Material | Potassium Phosphate Dibasic, Anhydrous |
Safety and Handling
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.