Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic, ACS (Dipotassium Hydrogen Phosphate), Reagents

Catalog No. 01337796
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-796 500 g
01-337-798 100 g
01-337-797 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-796 Supplier Reagents Holdings Llc Supplier No. C1289500500C2
Add to Cart
Add to Cart

Potassium Phosphate Dibasic, ACS (Dipotassium Hydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Phosphate Dibasic, ACS (Dipotassium Hydrogen Phosphate)
CAS 11-4-7758
Color Colorless/White
Physical Form Solid
Quantity 500 g
Molecular Formula K2HPO4
Synonym Dipotassium Hydrogen Phosphate
Molecular Weight (g/mol) 174.1759
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.