Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Dibasic, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCP065050F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCP065050F 50 kg Fiber Drum
RDCP0650500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCP065050F Supplier Ricca Chemical Company Supplier No. RDCP065050F4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Powder
Quantity 50 kg
Percent Purity ≥ 98%
Grade ACS
Packaging Fiber Drum
Chemical Name or Material Potassium Phosphate, Dibasic
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.