Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 10 Kg
SDP
Catalog No. 5000661339
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (10 kg), is a high-quality laboratory reagent designed for bulk applications requiring large quantities of potassium phosphate. This salt is essential for formulating buffers and solutions in biochemical and molecular biology experiments. As an anhydrous compound, it ensures precise control over solution concentrations by eliminating the need to account for water content. It is widely used in enzymology, microbiology, and plant biology, where potassium and phosphate ions are crucial for cellular processes, nutrient absorption, and buffer capacity. Its large volume makes it suitable for industrial-scale research and educational laboratories that require reliable, high-volume chemical reagents.
Catalog No. 50-006-61339 Supplier Alkali Scientific Supplier No. POA011CP
Potassium Phosphate Dibasic, Anhydrous (10 kg), is a high-quality laboratory reagent designed for bulk applications requiring large quantities of potassium phosphate. This salt is essential for formulating buffers and solutions in biochemical and molecular biology experiments. As an anhydrous compound, it ensures precise control over solution concentrations by eliminating the need to account for water content. It is widely used in enzymology, microbiology, and plant biology, where potassium and phosphate ions are crucial for cellular processes, nutrient absorption, and buffer capacity. Its large volume makes it suitable for industrial-scale research and educational laboratories that require reliable, high-volume chemical reagents.