Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 25 Kg
SDP
Catalog No. 5000660979
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (25 kg), is a bulk laboratory reagent designed for extensive use in chemical and biological experiments. This large volume is suitable for high-throughput laboratories and manufacturing facilities that require large quantities of potassium phosphate for buffer preparation, analytical chemistry, and other applications. As an anhydrous salt, it ensures that no moisture interferes with the preparation of precise solutions, making it ideal for experiments that demand exact concentrations of phosphate ions. Potassium Phosphate Dibasic is widely used in plant biology, microbial studies, and chemical processes where phosphate plays a pivotal role in nutrient provision and pH regulation.
Catalog No. 50-006-60979 Supplier Alkali Scientific Supplier No. POA014CP
Potassium Phosphate Dibasic, Anhydrous (25 kg), is a bulk laboratory reagent designed for extensive use in chemical and biological experiments. This large volume is suitable for high-throughput laboratories and manufacturing facilities that require large quantities of potassium phosphate for buffer preparation, analytical chemistry, and other applications. As an anhydrous salt, it ensures that no moisture interferes with the preparation of precise solutions, making it ideal for experiments that demand exact concentrations of phosphate ions. Potassium Phosphate Dibasic is widely used in plant biology, microbial studies, and chemical processes where phosphate plays a pivotal role in nutrient provision and pH regulation.