Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 500 G
SDP
Catalog No. 5000661212
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (500 g), is a high-purity salt that provides a reliable source of potassium and phosphate ions for smaller laboratory applications. This compact 500 g package is ideal for precise experiments and reagent formulations where larger quantities are not needed. It is widely used in the preparation of phosphate buffers, biological media, and as a nutrient source in plant and microbial research. The anhydrous form of potassium phosphate ensures accurate solution concentration without the interference of water content. This reagent is essential for experiments in enzymology, microbiology, and chemical analysis where phosphate plays a vital role in reactions and nutrient support.
Catalog No. 50-006-61212 Supplier Alkali Scientific Supplier No. POA016CP
Potassium Phosphate Dibasic, Anhydrous (500 g), is a high-purity salt that provides a reliable source of potassium and phosphate ions for smaller laboratory applications. This compact 500 g package is ideal for precise experiments and reagent formulations where larger quantities are not needed. It is widely used in the preparation of phosphate buffers, biological media, and as a nutrient source in plant and microbial research. The anhydrous form of potassium phosphate ensures accurate solution concentration without the interference of water content. This reagent is essential for experiments in enzymology, microbiology, and chemical analysis where phosphate plays a vital role in reactions and nutrient support.