Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, ACS, 3 Kg
SDP
Catalog No. 5000661230
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous, ACS (3 kg), is a high-purity chemical reagent that is perfect for advanced research and laboratory applications requiring reliable phosphate buffering. The ACS grade ensures high purity and consistency, making it ideal for use in pharmaceutical, chemical, and biochemical experiments. It is widely used for preparing phosphate buffer solutions, controlling pH levels, and providing potassium and phosphate ions for enzymatic reactions and microbial growth. This product is essential in laboratory settings that require high-quality reagents for precise experimental conditions and is commonly used in analytical chemistry, molecular biology, and biochemistry.
Catalog No. 50-006-61230 Supplier Alkali Scientific Supplier No. POA018CP
Potassium Phosphate Dibasic, Anhydrous, ACS (3 kg), is a high-purity chemical reagent that is perfect for advanced research and laboratory applications requiring reliable phosphate buffering. The ACS grade ensures high purity and consistency, making it ideal for use in pharmaceutical, chemical, and biochemical experiments. It is widely used for preparing phosphate buffer solutions, controlling pH levels, and providing potassium and phosphate ions for enzymatic reactions and microbial growth. This product is essential in laboratory settings that require high-quality reagents for precise experimental conditions and is commonly used in analytical chemistry, molecular biology, and biochemistry.