Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic, Anhydrous, BiotechGrade, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605663 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-663 500 g Poly Bottle
18-605-664 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-663 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1386500GM
Add to Cart
Add to Cart

P1386, 7758-11-4, K2HPO4

Spectrum™ Chemical Potassium Phosphate Dibasic, Anhydrous, BiotechGrade , also known as Potassium monohydrogen phosphate, is used to make buffer solutions. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

pH Approx. 9
CAS Max % 1
Quantity 500 g
Percent Purity 98.0 to 100.5%
Grade Biotech
Loss on Drying 0.01
Odor Odorless
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.