Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic, MP Biomedicals™

Catalog No. MP021519451 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021519451 100 g
MP021519455 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021519451 Supplier MP Biomedicals, Inc Supplier No. 02151945.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a component for a wide variety of media used in the culture of microorganisms and in phosphate buffered saline (PBS). Potassium Phosphate Dibasic, MP Biomedicals is a reagent with high buffering capacity. In addition to helping maintain pH, it sup

Potassium phosphate occurs in several forms: monobasic, dibasic and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water (1g/20mL); Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula Weight 174.18
Percent Purity ≥98% (Anhydrous)
Grade Not Available
Chemical Name or Material Potassium Phosphate, Dibasic
Recommended Storage Room Temperature(15–30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.