Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic, Purified (Dipotassium Hydrogen Phosphate), Reagents

Catalog No. 01337092
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-092 100 g
01-337-091 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-092 Supplier Reagents Holdings Llc Supplier No. C2289500100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Phosphate Dibasic, Purified (Dipotassium Hydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym Dipotassium Hydrogen Phosphate
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 g
Percent Purity 98%
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.3
Chemical Name or Material Potassium Phosphate Dibasic, Purified (Dipotassium Hydrogen Phosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.